Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02765420_gH
          See other GM6498 GE Assays ›
          Gene Symbol:
          Gm6498
          Gene Name:
          glyceraldehyde-3-phosphate dehydrogenase pseudogene
          Gene Aliases:
          EG624367
          Chromosome Location:
          Chr.14: 48198716 - 48228413 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02765420_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm6498
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003630.2 3 1482 61
          GenBank mRNA AK033118.1 - 3 1482 61

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          dehydrogenase

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P291460 NR_003630.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P291460 AK033118.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P291460 NR_003630.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P291460 AK033118.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P291460 NR_003630.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P291460 AK033118.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P291460 NR_003630.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P291460 AK033118.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P291460 NR_003630.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P291460 AK033118.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P291460 NR_003630.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P291460 AK033118.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140564 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10414429 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414428 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414430 - -
          Affymetrix Mouse Gene 1.1 ST Array 10414427 - -
          Affymetrix Mouse Clariom S Array PSR1400004282.mm.1 NR_003630.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004282.mm.1 TC1400000528.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400004281.mm.1 NR_003630.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004281.mm.1 TC1400000528.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400004276.mm.1 NR_003630.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004276.mm.1 TC1400000528.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400004280.mm.1 NR_003630.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004280.mm.1 TC1400000528.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400004277.mm.1 NR_003630.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004277.mm.1 TC1400000528.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 395587 - -
          Array Name Probe IDs Targets Target Type
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2960286 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline