Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03011781_m1
          See other KIFC5B GE Assays ›
          Gene Symbol:
          Kifc5b Kifc1
          Gene Name:
          kinesin family member C5B
          kinesin family member C1
          Gene Aliases:
          Hset, KNSL2, Kifc5a
          100042970, Gm4137, HSET, KNSL2, Kifc5a, Knsl2a, Tctex-7, Tctex-7A, Tctex7, Tctex7a
          Chromosome Location:
          Chr.17: 26917091 - 26932579 on Build GRCm38
          Chromosome Location:
          Chr.17: 33875664 - 33890633 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm03011781_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kifc5b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053173.2 NP_444403.2 9 - 10 2074 101
          GenBank mRNA AK081484.1 - 3 - 4 745 101
          Gene Symbol: Kifc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001195298.1 NP_001182227.1 9 - 10 2041 101
          GenBank mRNA AF221102.1 - 9 - 10 2024 101
          AI326799.1 - 2 - 3 101
          AK133900.1 - 9 - 10 2047 101
          AM051187.1 - 8 - 9 1774 101
          BC003753.1 - 9 - 10 1952 3495308 101
          BC057162.1 - 9 - 10 1977 5350661 101
          BC100328.1 - 9 - 10 1996 30841951 101
          D49544.1 - 10 - 11 1761 101
          KJ650206.1 - 9 - 10 1903 101

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kifc5b
          Click here to find siRNAs for Gene Kifc1

          Panther Classification:

          Molecular Function -

          microtubule binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic sister chromatid segregation
          microtubule-based movement
          spindle assembly
          negative regulation of centrosome duplication
          vesicle transport along microtubule
          minus-end-directed vesicle transport along microtubule
          ATP-dependent microtubule motor activity, minus-end-directed
          microtubule motor activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113187 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113143 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113145 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113146 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1987028 AI604818.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1987028 NM_053173.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2062543 BC003753.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2062543 BC057162.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2062543 BC100328.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2062543 AI326799.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2062543 AK133900.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 809862 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 277123 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 144518 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153866 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1987028 AI604818.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1987028 NM_053173.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113143 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08746667 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153866 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08746658 - -
          Whole Mouse Genome G4122A A_51_P137433 D49544.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P137433 BC003753.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P137433 BC057162.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P137433 BC100328.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P137433 AI326799.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P550734 BC003753.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P550734 BC057162.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P550734 BC100328.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P550734 AI326799.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P550734 AK133900.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08746667 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153866 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13468817 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13207389 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13207390 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13207396 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113143 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113146 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113145 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13207390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13207393 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13158239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13130103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13105030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13130105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13207389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13421212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13131262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13432215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13372330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13087567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113227 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10443056 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443054 - -
          Affymetrix Mouse Gene 1.0 ST Array 10366545 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443048 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443062 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443060 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443053 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443049 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443061 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443052 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443059 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443050 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443051 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443055 - -
          Affymetrix HT MG-430 PM Array Plate 1456136_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10366544 - -
          Affymetrix Mouse Gene 1.1 ST Array 10443047 - -
          Mouse Genome 430 2.0 Array 1456136_at - -
          Mouse Genome 430 2.0 Array 1449877_s_at - -
          Mouse Genome 430 2.0 Array 1449877_PM_s_at - -
          Mouse Genome 430 2.0 Array 1456136_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700017452.mm.1 NM_001195298.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017452.mm.1 NM_001195298.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017452.mm.1 TC1700001881.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700003645.mm.1 NM_053173.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003645.mm.1 NM_053173.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003645.mm.1 TC1700000424.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700003646.mm.1 NM_053173.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003646.mm.1 NM_053173.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003646.mm.1 TC1700000424.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700017451.mm.1 NM_001195298.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017451.mm.1 NM_001195298.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017451.mm.1 TC1700001881.mm.1 MTA TC
          Mouse Expression Set 430 1449877_s_at - -
          Mouse Expression Set 430 1449877_PM_s_at - -
          Murine Genome U74v2 Set 98471_PM_f_at - -
          Murine Genome U74v2 Set 98471_f_at - -
          Murine Genome U74v2 Set 169214_PM_at - -
          Murine Genome U74v2 Set 169214_at - -
          Mouse Genome 430A 2.0 Array 1449877_s_at - -
          Mouse Genome 430A 2.0 Array 1449877_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1456136_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1456136_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 784383 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6840021 - -
          Illumina mouse-6 v1.0 expression beadchip 6840021 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006840021 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1231176 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231176 - -
          mouse-6 v1.1 (Illumina) scl0094116.1_13-S - -
          Illumina Sentrix mouse-6.v1 bead array 6840021 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0094116.1_13-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6840021 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226979 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline