Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03012639_s1
          See other OLFR50 GE Assays ›
          Gene Symbol:
          Olfr50
          Gene Name:
          olfactory receptor 50
          Gene Aliases:
          ID3, MOR136-6
          Chromosome Location:
          Chr.2: 36793238 - 36794176 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03012639_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr50
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146946.1 NP_667157.1 1 554 143
          GenBank mRNA AF102524.1 - 1 365 143
          BC127978.1 - 1 558 40135208 143

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr50

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          response to stimulus
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          signal transducer activity
          G-protein coupled receptor activity
          olfactory receptor activity
          isoprenoid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989623 NM_146946.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989623 BC127978.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2145481 NM_146946.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2145481 BC127978.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153697 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2145481 NM_146946.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2145481 BC127978.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989623 NM_146946.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989623 BC127978.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P334449 NM_146946.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P334449 AF102524.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P334449 BC127978.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21140022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21140008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21140000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21140004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21140020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21140006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21140018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21140002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21140017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21140013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21140019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21140014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21140016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21140009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21140003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21140023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139991 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153697 - -
          Whole Mouse Genome G4122A A_51_P334449 NM_146946.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334449 AF102524.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334449 BC127978.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111737 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153697 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01259315 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01259308 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01259312 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01259311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21140017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01883975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01896316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140046 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01984236 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01287265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02024727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01984237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139995 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140003 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140030 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01564260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21140050 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10471758 - -
          Affymetrix Mouse Gene 1.1 ST Array 10471757 - -
          Affymetrix Mouse Clariom S Array PSR0200006036.mm.1 NM_146946.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006036.mm.1 NM_146946.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006036.mm.1 TC0200000669.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 808273 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3830288 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1255688 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255688 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1255688 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline