Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03012894_g1
          See other TUFM GE Assays ›
          Gene Symbol:
          Tufm
          Gene Name:
          Tu translation elongation factor, mitochondrial
          Gene Aliases:
          2300002G02Rik, C76308, C76389, EF-TuMT, EFTU
          Chromosome Location:
          Chr.7: 126487265 - 126490731 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03012894_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tufm
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163713.1 NP_001157185.1 8 - 9 1369 85
          NM_172745.3 NP_766333.1 9 - 10 1281 85
          GenBank mRNA AK075857.1 - 9 - 10 1205 85
          AK084724.1 - 9 - 10 1262 85
          AK152858.1 - 9 - 10 1276 85
          AK153135.1 - 9 - 10 1276 85
          BC060959.1 - 8 - 9 1346 30379757 85
          BC100596.1 - 9 - 10 1274 6762419 85

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tufm

          Panther Classification:

          Molecular Function -

          translation elongation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          translational elongation
          nucleotide binding
          translation elongation factor activity
          GTPase activity
          GTP binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25500777 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25500776 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2140711 AK208201.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2140711 AK153135.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2140711 AK195139.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2140711 BC060959.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2140711 BC100596.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 482265 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865212 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 939856 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142808 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2151021 BC100596.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2140711 AK208201.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2140711 AK153135.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2140711 AK195139.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2140711 BC060959.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2140711 BC100596.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P165617 AK208201.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P165617 AK153135.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P165617 AK195139.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P165617 BC060959.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P165617 BC100596.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500763 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142808 - -
          Whole Mouse Genome G4122A A_51_P165617 AK208201.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165617 AK153135.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165617 AK195139.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165617 BC060959.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165617 BC100596.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113440 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140243 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142808 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06244518 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06244506 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06244535 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06026868 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06244507 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25500776 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25500777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06244506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06004786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06004783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06004785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06244503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06244527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25500795 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10557429 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557421 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557426 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557431 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557424 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578985 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557427 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557422 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557428 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557430 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557425 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557423 - -
          Affymetrix HT MG-430 PM Array Plate 1449673_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1447780_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1433952_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1449672_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10557420 - -
          Affymetrix Mouse Gene 1.1 ST Array 10578984 - -
          Mouse Genome 430 2.0 Array 1447780_PM_x_at - -
          Mouse Genome 430 2.0 Array 1449673_PM_s_at - -
          Mouse Genome 430 2.0 Array 1449672_PM_s_at - -
          Mouse Genome 430 2.0 Array 1449673_s_at - -
          Mouse Genome 430 2.0 Array 1433952_PM_at - -
          Mouse Genome 430 2.0 Array 1449672_s_at - -
          Mouse Genome 430 2.0 Array 1447780_x_at - -
          Mouse Genome 430 2.0 Array 1433952_at - -
          Affymetrix Mouse Clariom S Array PSR0700015385.mm.1 NM_001163713.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015385.mm.1 NM_001163713.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015385.mm.1 NM_172745.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015385.mm.1 NM_172745.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015385.mm.1 TC0700001804.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700015386.mm.1 NM_001163713.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015386.mm.1 NM_001163713.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015386.mm.1 NM_172745.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015386.mm.1 NM_172745.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015386.mm.1 TC0700001804.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700015384.mm.1 NM_001163713.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015384.mm.1 NM_001163713.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015384.mm.1 NM_172745.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015384.mm.1 NM_172745.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700015384.mm.1 TC0700001804.mm.1 MTA TC
          Mouse Expression Set 430 1433952_PM_at - -
          Mouse Expression Set 430 1433952_at - -
          Mouse Expression Set 430 1447780_x_at - -
          Mouse Expression Set 430 1447780_PM_x_at - -
          Murine Genome U74v2 Set 161120_r_at - -
          Murine Genome U74v2 Set 96626_PM_at - -
          Murine Genome U74v2 Set 96626_at - -
          Murine Genome U74v2 Set 161120_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1433952_PM_at - -
          Mouse Genome 430A 2.0 Array 1433952_at - -
          Affymetrix HT Mouse Genome 430A Array 1449673_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1433952_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449672_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1433952_at - -
          Affymetrix HT Mouse Genome 430A Array 1449672_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1449673_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1447780_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1447780_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 903158 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3710039 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003710039 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2462448 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2462448 - -
          mouse-6 v1.1 (Illumina) scl0233870.10_246-S - -
          Illumina Sentrix mouse-6.v1 bead array 3710039 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0233870.10_246-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3710039 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2462448 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2462448 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0233870.10_246-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline