Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03030107_s1
          See other EVI2A GE Assays ›
          Gene Symbol:
          Evi2a
          Gene Name:
          ecotropic viral integration site 2a
          Gene Aliases:
          AW491894, Evi-2, Evi2
          Chromosome Location:
          Chr.11: 79526560 - 79530609 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03030107_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Evi2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033711.1 NP_001028883.1 2 1570 116
          NM_010161.3 NP_034291.1 2 757 116
          GenBank mRNA AK044098.1 - 2 633 116
          AK046407.1 - 2 708 116
          AK160302.1 - 2 674 116
          AK162958.1 - 633 116
          AK170527.1 - 2 729 116
          AK171332.1 - 1 3114 116
          AK171594.1 - 2 675 116
          BC038124.1 - 2 714 1396641 116
          M34896.1 - 2 687 116
          M34897.1 - 1 1083 116

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Evi2a

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017759 BC038124.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017759 AK170527.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017759 AK171594.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017759 AK160302.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017759 AK162958.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112004 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177193 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017759 BC038124.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017759 AK170527.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017759 AK171594.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017759 AK160302.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017759 AK162958.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949399 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949410 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949395 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949397 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949403 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949405 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27949409 - -
          Whole Mouse Genome G4122A A_52_P380895 BC038124.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P380895 M34896.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P380895 M34897.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P380895 AK170527.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P380895 AK171594.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103675 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09373569 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09373571 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09373575 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09381574 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09373578 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09373573 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09373577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27949461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09065498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09373570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09429025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09411888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27949455 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10388967 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388968 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388966 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388963 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388964 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388965 - -
          Affymetrix HT MG-430 PM Array Plate 1443894_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1450241_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10388958 - -
          Mouse Genome 430 2.0 Array 1426506_at - -
          Mouse Genome 430 2.0 Array 1443894_PM_at - -
          Mouse Genome 430 2.0 Array 1443894_at - -
          Mouse Genome 430 2.0 Array 1426506_PM_at - -
          Mouse Genome 430 2.0 Array 1450241_PM_a_at - -
          Mouse Genome 430 2.0 Array 1443097_at - -
          Mouse Genome 430 2.0 Array 1443097_PM_at - -
          Mouse Genome 430 2.0 Array 1450241_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14017_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14017_at - -
          Affymetrix Mouse Clariom S Array PSR1100030605.mm.1 NM_001033711.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030605.mm.1 NM_001033711.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030605.mm.1 NM_010161.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030605.mm.1 NM_010161.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030605.mm.1 TC1100003326.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100030607.mm.1 NM_001033711.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030607.mm.1 NM_001033711.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030607.mm.1 NM_010161.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030607.mm.1 NM_010161.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030607.mm.1 TC1100003326.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100030606.mm.1 NM_001033711.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030606.mm.1 NM_001033711.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030606.mm.1 NM_010161.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030606.mm.1 NM_010161.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030606.mm.1 TC1100003326.mm.1 MTA TC
          Mouse Expression Set 430 1450241_PM_a_at - -
          Mouse Expression Set 430 1426506_PM_at - -
          Mouse Expression Set 430 1450241_a_at - -
          Mouse Expression Set 430 1426506_at - -
          Mouse Expression Set 430 1443894_PM_at - -
          Mouse Expression Set 430 1443894_at - -
          Murine Genome U74v2 Set 98026_PM_g_at - -
          Murine Genome U74v2 Set 98025_at - -
          Murine Genome U74v2 Set 98026_g_at - -
          Murine Genome U74v2 Set 98025_PM_at - -
          Mouse Genome 430A 2.0 Array 1450241_a_at - -
          Mouse Genome 430A 2.0 Array 1426506_PM_at - -
          Mouse Genome 430A 2.0 Array 1426506_at - -
          Mouse Genome 430A 2.0 Array 1450241_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1450241_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1450241_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1443894_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443894_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6840110 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006840110 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1251669 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1251669 - -
          mouse-6 v1.1 (Illumina) scl0014017.1_112-S - -
          Illumina Sentrix mouse-6.v1 bead array 6840110 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0014017.1_112-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6840110 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3153285 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1251669 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3153285 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1251669 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0014017.1_112-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline