Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03031105_s1
          See other CD164L2 GE Assays ›
          Gene Symbol:
          Cd164l2
          Gene Name:
          CD164 sialomucin-like 2
          Gene Aliases:
          2310047N01Rik
          Chromosome Location:
          Chr.4: 133220809 - 133224554 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03031105_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd164l2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027152.1 NP_081428.1 2 347 86
          GenBank mRNA AK009888.1 - 2 273 86
          AK139809.1 - 2 348 86

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23316407 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23316408 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2181904 NM_027152.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2181904 AK009888.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2181904 AK139809.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964440 NM_027152.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964440 AK009888.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964440 AK139809.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964440 BC111824.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 214444 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964440 NM_027152.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964440 AK009888.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964440 AK139809.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964440 BC111824.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2181904 NM_027152.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2181904 AK009888.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2181904 AK139809.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P354757 NM_027152.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P354757 AK009888.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P354757 AK139809.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P354757 BC111824.1 mouse_transcriptome
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23316407 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23316408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23316361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03809822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03824011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03280438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03974172 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03291176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03280447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03974171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03291179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03204870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03809825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04043794 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03280436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04043791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316423 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03280426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03291177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03280453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03291175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03291181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03530640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03291182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03948031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03280423 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03204871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316406 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03280451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03889525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03280435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23316425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03672123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03280433 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10508821 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508822 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508824 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508826 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508827 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508823 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508828 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508825 - -
          Affymetrix HT MG-430 PM Array Plate 1429602_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1443664_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10508819 - -
          Mouse Genome 430 2.0 Array 1429602_PM_at - -
          Mouse Genome 430 2.0 Array 1443664_PM_s_at - -
          Mouse Genome 430 2.0 Array 1429602_at - -
          Mouse Genome 430 2.0 Array 1443664_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69655_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69655_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400012685.mm.1 NM_027152.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012685.mm.1 NM_027152.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012685.mm.1 TC0400001606.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400012674.mm.1 NM_027152.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012674.mm.1 NM_027152.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012674.mm.1 TC0400001606.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400012683.mm.1 NM_027152.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012683.mm.1 NM_027152.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012683.mm.1 TC0400001606.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400012676.mm.1 NM_027152.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012676.mm.1 NM_027152.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012676.mm.1 TC0400001606.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400012673.mm.1 NM_027152.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012673.mm.1 NM_027152.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012673.mm.1 TC0400001606.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400012677.mm.1 NM_027152.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012677.mm.1 NM_027152.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012677.mm.1 TC0400001606.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400012678.mm.1 NM_027152.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012678.mm.1 NM_027152.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012678.mm.1 TC0400001606.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400012680.mm.1 NM_027152.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012680.mm.1 NM_027152.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012680.mm.1 TC0400001606.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400012681.mm.1 NM_027152.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012681.mm.1 NM_027152.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012681.mm.1 TC0400001606.mm.1 MTA TC
          Mouse Expression Set 430 1429602_PM_at - -
          Mouse Expression Set 430 1429602_at - -
          Mouse Expression Set 430 1443664_PM_s_at - -
          Mouse Expression Set 430 1443664_s_at - -
          Murine Genome U74v2 Set 169600_PM_at - -
          Murine Genome U74v2 Set 130971_at - -
          Murine Genome U74v2 Set 130971_PM_at - -
          Murine Genome U74v2 Set 169600_at - -
          Affymetrix HT Mouse Genome 430B Array 1429602_at - -
          Affymetrix HT Mouse Genome 430B Array 1443664_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1429602_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443664_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2617449 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2617449 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline