Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03031714_s1
          See other TCEAL1 GE Assays ›
          Gene Symbol:
          Tceal1
          Gene Name:
          transcription elongation factor A (SII)-like 1
          Gene Aliases:
          0610011M09Rik, BC011290, P21, SIIR, pp21
          Chromosome Location:
          Chr.X: 136707966 - 136709873 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03031714_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tceal1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146236.1 NP_666348.1 2 459 139
          GenBank mRNA BC011290.1 - 2 459 4010035 139
          BC083157.1 - 2 493 5376199 139

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tceal1

          Panther Classification:

          Molecular Function -

          general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          biological_process
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P249930 NM_146236.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P249930 BC011290.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P249930 BC083157.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P249930 NM_146236.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P249930 BC011290.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P249930 BC083157.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 153710 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153509 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P249930 NM_146236.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P249930 BC011290.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P249930 BC083157.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P249930 NM_146236.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P249930 BC011290.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P249930 BC083157.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249930 NM_146236.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249930 BC011290.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249930 BC083157.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249930 NM_146236.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249930 BC011290.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P249930 BC083157.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32681974 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153509 - -
          Whole Mouse Genome G4122A A_51_P249930 NM_146236.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249930 BC011290.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249930 BC083157.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249930 NM_146236.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249930 BC011290.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249930 BC083157.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102070 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153509 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15037808 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15037818 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14837036 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15037802 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15037804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32681999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14683552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14837026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15037804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15200279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14560253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14575221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14619556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14599468 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14619559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14619570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32681991 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10601879 - -
          Affymetrix HT MG-430 PM Array Plate 1424634_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10601878 - -
          Mouse Genome 430 2.0 Array 1424634_at - -
          Mouse Genome 430 2.0 Array 1424634_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 237052_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 237052_at - -
          Affymetrix Mouse Clariom S Array PSR0X00008390.mm.1 TC0X00001335.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00008392.mm.1 NM_146236.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008392.mm.1 NM_146236.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008392.mm.1 TC0X00001335.mm.1 MTA TC
          Mouse Expression Set 430 1424634_at - -
          Mouse Expression Set 430 1424634_PM_at - -
          Mouse Genome 430A 2.0 Array 1424634_at - -
          Mouse Genome 430A 2.0 Array 1424634_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424634_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424634_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 569326 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 870577 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000870577 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2695731 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2695731 - -
          mouse-6 v1.1 (Illumina) scl54611.2_16-S - -
          Illumina Sentrix mouse-6.v1 bead array 870577 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl54611.2_16-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 870577 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2937136 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2695731 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2937136 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2695731 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl54611.2_16-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline