Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03039586_g1
          See other VMN1R171 GE Assays ›
          Gene Symbol:
          Vmn1r171 Vmn1r169 Vmn1r170 Vmn1r175 Vmn1r176
          Gene Name:
          vomeronasal 1 receptor 171
          vomeronasal 1 receptor 169
          vomeronasal 1 receptor 170
          vomeronasal 1 receptor 175
          vomeronasal 1 receptor 176
          Gene Aliases:
          V1rd7, V3R7
          Gm4232
          EG546944, Gm5999
          EG622222, Gm6299
          Gm5998
          Chromosome Location:
          Chr.7: 23631988 - 23633568 on Build GRCm38
          Chromosome Location:
          Chr.7: 23577185 - 23578099 on Build GRCm38
          Chromosome Location:
          Chr.7: 23606175 - 23607089 on Build GRCm38
          Chromosome Location:
          Chr.7: 23808286 - 23809200 on Build GRCm38
          Chromosome Location:
          Chr.7: 23834812 - 23835726 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          Assay ID Mm03039586_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r171
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030737.2 NP_109662.1 1 972 65
          GenBank mRNA AF324871.1 - 982 65
          BC111911.1 - 1 972 40046770 65
          Gene Symbol: Vmn1r169
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001166843.1 NP_001160315.1 1 572 65
          Gene Symbol: Vmn1r170
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001166722.1 NP_001160194.1 1 572 65
          Gene Symbol: Vmn1r175
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001166727.1 NP_001160199.1 1 572 65
          Gene Symbol: Vmn1r176
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001166721.1 NP_001160193.1 1 572 65

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r171
          Click here to find siRNAs for Gene Vmn1r169
          Click here to find siRNAs for Gene Vmn1r170
          Click here to find siRNAs for Gene Vmn1r175
          Click here to find siRNAs for Gene Vmn1r176

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988669 AF324871.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988669 BC111911.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988669 NM_030737.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2026913 NM_001166843.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1953292 NM_001166721.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106190 NM_001166727.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988669 AF324871.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988669 BC111911.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988669 NM_030737.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993450 NM_001166722.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P145988 AF324871.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P145988 BC111911.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P145988 NM_030737.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25002192 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06139538 - -
          Whole Mouse Genome G4122A A_51_P145988 AF324871.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P145988 BC111911.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P145988 NM_030737.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25002195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06222642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06367341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25002185 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10550814 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550813 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550812 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560811 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550811 - -
          Affymetrix HT MG-430 PM Array Plate 1421724_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10550810 - -
          Affymetrix Mouse Gene 1.1 ST Array 10550807 - -
          Affymetrix Mouse Gene 1.1 ST Array 10560810 - -
          Affymetrix Mouse Gene 1.1 ST Array 10560812 - -
          Affymetrix Mouse Gene 1.1 ST Array 10560810 - -
          Affymetrix Mouse Gene 1.1 ST Array 10560810 - -
          Affymetrix Mouse Gene 1.1 ST Array 10560810 - -
          Affymetrix Mouse Gene 1.1 ST Array 10560810 - -
          Mouse Genome 430 2.0 Array 1421724_at - -
          Mouse Genome 430 2.0 Array 1421724_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700002618.mm.1 TC0700000355.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002618.mm.1 NM_001166722.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002618.mm.1 NM_001166722.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002617.mm.1 TC0700000354.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002617.mm.1 NM_001166843.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002617.mm.1 NM_001166843.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700022354.mm.1 TC0700002585.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700022354.mm.1 NM_001166727.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700022354.mm.1 NM_001166727.1 Refseq
          Mouse Expression Set 430 1421724_PM_at - -
          Mouse Expression Set 430 1421724_at - -
          Mouse Genome 430A 2.0 Array 1421724_PM_at - -
          Mouse Genome 430A 2.0 Array 1421724_at - -
          Affymetrix HT Mouse Genome 430A Array 1421724_at - -
          Affymetrix HT Mouse Genome 430A Array 1421724_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 441328 - -
          ABI Mouse Genome Survey Microarray 484205 - -
          ABI Mouse Genome Survey Microarray 713781 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1780397 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001780397 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2446884 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2446884 - -
          mouse-6 v1.1 (Illumina) scl081012.3_170-S - -
          Illumina Sentrix mouse-6.v1 bead array 1780397 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl081012.3_170-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1780397 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2817661 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2446884 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2817661 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline