Search
Search

Akap2| Gene Symbol: Akap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035533.2 | NP_001030610.1 | 2 - 3 | 2660 | 66 | |
| GenBank mRNA | AF033274.1 | - | 2 - 3 | 3062 | 66 | |
| AW489512.1 | - | 2 - 3 | 310 | 66 | ||
Set Membership: |
|