Search
Search

Efcc1| Gene Symbol: Efcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159697.1 | NP_001153169.1 | 1 - 2 | 666 | 125 | |
| GenBank mRNA | AB041550.1 | - | 1 - 2 | 666 | 125 | |
| BC139438.1 | - | 1 - 2 | 663 | 8862460 | 125 | |
| BC139439.1 | - | 1 - 2 | 662 | 8862461 | 125 | |
Set Membership: |
|