| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700036361.mm.1 |
NM_175163.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036361.mm.1 |
NM_175163.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036361.mm.1 |
TC0700004328.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700036362.mm.1 |
NM_175163.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036362.mm.1 |
NM_175163.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036362.mm.1 |
TC0700004328.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700036360.mm.1 |
NM_175163.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036360.mm.1 |
NM_175163.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036360.mm.1 |
TC0700004328.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700036363.mm.1 |
NM_175163.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036363.mm.1 |
NM_175163.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700036363.mm.1 |
TC0700004328.mm.1 |
MTA TC |