Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03053870_g1
          See other MIXL1 GE Assays ›
          Gene Symbol:
          Mixl1
          Gene Name:
          Mix1 homeobox-like 1 (Xenopus laevis)
          Gene Aliases:
          Mm1, Mml
          Chromosome Location:
          Chr.1: 180693043 - 180709055 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03053870_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mixl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013729.3 NP_038757.1 2 2238 83
          GenBank mRNA AF154573.1 - 2 2239 83
          AK138043.1 - 1 1554 83

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mixl1

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          endoderm formation
          hematopoietic progenitor cell differentiation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          multicellular organism development
          gastrulation
          endoderm development
          heart development
          hemopoiesis
          cell differentiation
          endodermal cell differentiation
          cell migration involved in gastrulation
          positive regulation of transcription from RNA polymerase II promoter
          digestive tract development
          negative regulation of hematopoietic progenitor cell differentiation
          positive regulation of mesoderm development
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription factor binding
          RNA polymerase II repressing transcription factor binding
          transcriptional activator activity, RNA polymerase II transcription factor binding
          transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
          DNA binding
          protein homodimerization activity
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20877314 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20877308 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20877313 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20877310 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20877309 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20877312 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20877311 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P533129 AF201959.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P533129 AK138043.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P533129 NM_013729.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P533129 AF154573.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P533129 AF201959.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P533129 AK138043.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P533129 NM_013729.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P533129 AF154573.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 423978 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 893511 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 185167 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152368 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P533129 AF201959.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P533129 AK138043.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P533129 NM_013729.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P533129 AF154573.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P533129 AF201959.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P533129 AK138043.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P533129 NM_013729.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P533129 AF154573.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20877322 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152368 - -
          Whole Mouse Genome G4122A A_52_P533129 AF201959.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P533129 AK138043.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P533129 NM_013729.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P533129 AF154573.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P533129 AF201959.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P533129 AK138043.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P533129 NM_013729.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P533129 AF154573.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125542 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152368 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01062584 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01062577 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01062583 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01062569 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20877308 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20877310 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20877314 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20877311 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20877312 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20877313 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20877309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20877290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00951128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00951124 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01062574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00911993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00951134 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00165648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00738789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00346856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00451958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00346852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00325507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00346855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00556472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00951135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00165644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01062577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01062587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00183744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20877298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01019256 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10360677 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360678 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360675 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360676 - -
          Affymetrix HT MG-430 PM Array Plate 1421505_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10360674 - -
          Mouse Genome 430 2.0 Array 1421505_PM_at - -
          Mouse Genome 430 2.0 Array 1421505_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 27217_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 27217_PM_at - -
          Mouse Expression Set 430 1421505_at - -
          Mouse Expression Set 430 1421505_PM_at - -
          Mouse Genome 430A 2.0 Array 1421505_PM_at - -
          Mouse Genome 430A 2.0 Array 1421505_at - -
          Affymetrix HT Mouse Genome 430A Array 1421505_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421505_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 359339 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1940017 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001940017 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245893 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245893 - -
          mouse-6 v1.1 (Illumina) scl15843.2_54-S - -
          Illumina Sentrix mouse-6.v1 bead array 1940017 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl15843.2_54-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1940017 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1245893 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2920173 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1245893 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2920173 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline