Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03412454_m1
          See other CYP26C1 GE Assays ›
          Gene Symbol:
          Cyp26c1
          Gene Name:
          cytochrome P450, family 26, subfamily c, polypeptide 1
          Gene Aliases:
          EG546726
          Chromosome Location:
          Chr.19: 37685680 - 37693307 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03412454_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp26c1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105201.1 NP_001098671.1 3 - 4 709 75
          GenBank mRNA BC151098.1 - 3 - 4 808 9088011 75
          BC151106.1 - 3 - 4 808 9088019 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000537.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          central nervous system development
          anterior/posterior pattern specification
          neural crest cell development
          retinoic acid catabolic process
          organelle fusion
          oxidation-reduction process
          retinoic acid binding
          retinoic acid 4-hydroxylase activity

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035407 NM_001105201.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035407 BC151098.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035407 BC151106.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 861371 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 907220 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 250250 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 409882 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 65951 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035407 NM_001105201.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035407 BC151098.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035407 BC151106.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32096983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32096974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32096965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32096977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32096991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32096989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32096995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32096976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32096984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32096967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32096972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32096970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32096997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32096996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32096980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32096981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32096985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32097013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14147522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14147496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32096975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32097008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14147547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14147508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32096984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32097015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32097036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32096968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32097003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32097041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32096997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32097044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14331181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14506651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32096972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14209603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14506652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32096976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32096989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14506653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32096974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32097012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32096967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32097017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32097039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32097006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32096996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32097042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32097019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32097029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32097033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14331165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14506661 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32096970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32097009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14147498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32096981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32096965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14147519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32096991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32097047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14147520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32097037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14226126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32096985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14331157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32097018 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10462852 - -
          Affymetrix Mouse Gene 1.0 ST Array 10462848 - -
          Affymetrix Mouse Gene 1.0 ST Array 10462849 - -
          Affymetrix Mouse Gene 1.0 ST Array 10462851 - -
          Affymetrix Mouse Gene 1.0 ST Array 10462850 - -
          Affymetrix Mouse Gene 1.0 ST Array 10462847 - -
          Affymetrix Mouse Gene 1.1 ST Array 10462846 - -
          Affymetrix Mouse Clariom S Array PSR1900004820.mm.1 NM_001105201.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004820.mm.1 NM_001105201.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004820.mm.1 TC1900000537.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900004822.mm.1 NM_001105201.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004822.mm.1 NM_001105201.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004822.mm.1 TC1900000537.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900004819.mm.1 NM_001105201.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004819.mm.1 NM_001105201.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004819.mm.1 TC1900000537.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900004821.mm.1 NM_001105201.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004821.mm.1 NM_001105201.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004821.mm.1 TC1900000537.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900004818.mm.1 TC1900000537.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline