| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700017847.mm.1 |
NM_030746.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017847.mm.1 |
NM_030746.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017847.mm.1 |
TC1700001909.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017846.mm.1 |
NM_030746.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017846.mm.1 |
NM_030746.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017846.mm.1 |
TC1700001909.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017849.mm.1 |
NM_030746.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017849.mm.1 |
NM_030746.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017849.mm.1 |
TC1700001909.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017848.mm.1 |
NM_030746.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017848.mm.1 |
NM_030746.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017848.mm.1 |
TC1700001909.mm.1 |
MTA TC |