Search
Search
| Gene Symbol: Tsix | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002844.2 | 2 - 3 | 514 | 72 | |||
| GenBank mRNA | AF138745.1 | - | LIT1604 Tsix |
2 - 3 | 514 | 72 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | testis specific X-linked gene | Tsx | AV207184 | -23.1kb | Sense | Chr. X - 103409101 - 103424588 on NCBI Build GRCm38 |
| Downstream | inactive X specific transcripts | Xist |
A430022B11, AI314753 |
12.1kb | Anti-sense | Chr. X - 103460373 - 103483233 on NCBI Build GRCm38 |
Set Membership: |
|