Search
Search

Peg13| Gene Symbol: Peg13 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002864.1 | 1 | 4480 | 72 | |||
| GenBank mRNA | AY151252.1 | - | LIT1829 Peg13 |
1 | 4480 | 72 | |
| AY151253.1 | - | LIT1828 Peg13 |
1 | 4154 | 72 | ||
Set Membership: |
|