Search
Search

Sec14l5| Gene Symbol: Sec14l5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127725.1 | NP_001121197.1 | 4 - 5 | 403 | 68 | |
| GenBank mRNA | BC157907.1 | - | 4 - 5 | 469 | 9088109 | 68 |
| BC157990.1 | - | 4 - 5 | 403 | 9007304 | 68 | |
Set Membership: |
|