Search
Search

Fyttd1| Gene Symbol: Fyttd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159349.1 | NP_001152821.1 | 2 - 3 | 345 | 102 | |
| NM_027226.4 | NP_081502.2 | 2 - 3 | 345 | 102 | ||
| GenBank mRNA | AJ344096.1 | - | 2 - 3 | 229 | 102 | |
| AK008130.1 | - | 2 - 3 | 317 | 102 | ||
| AK014447.1 | - | 2 - 3 | 316 | 102 | ||
| AK029785.1 | - | 2 - 3 | 290 | 102 | ||
| AK050448.1 | - | 2 - 3 | 304 | 102 | ||
| AK050479.1 | - | 2 - 3 | 317 | 102 | ||
| AK139638.1 | - | 2 - 3 | 341 | 102 | ||
| AK157602.1 | - | 2 - 3 | 317 | 102 | ||
| AK161252.1 | - | 2 - 3 | 323 | 102 | ||
| AK165633.1 | - | 2 - 3 | 299 | 102 | ||
| AK166032.1 | - | 2 - 3 | 317 | 102 | ||
| AK170511.1 | - | 2 - 3 | 317 | 102 | ||
| BC010204.1 | - | 2 - 3 | 319 | 3158224 | 102 | |
| BC028253.1 | - | 2 - 3 | 302 | 3983533 | 102 | |
Set Membership: |
|