Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03808897_g1
          See other TEDDM3 GE Assays ›
          Gene Symbol:
          Teddm3
          Gene Name:
          transmembrane epididymal family member 3
          Gene Aliases:
          2310042E22Rik
          Chromosome Location:
          Chr.16: 21152659 - 21153944 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03808897_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Teddm3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025634.3 NP_079910.1 1 754 107
          GenBank mRNA AK009367.1 - 1 698 107
          AK009496.1 - 1 712 106
          AK009566.1 - 1 756 107
          AK009632.1 - 1 715 107
          AK009635.1 - 1 716 107
          AK009751.1 - 1 813 107
          AK009766.1 - 1 706 107
          AK009777.1 - 1 698 107
          AK009984.1 - 1 704 107
          AK009997.1 - 1 750 107
          AK010016.1 - 1 715 107
          BC115655.1 - 1 705 40103183 107
          BC115656.1 - 1 704 40103184 107

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Teddm3

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994418 AK009984.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994418 AK009997.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994418 AK010016.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994418 AK009566.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994418 AK009632.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994418 AK009984.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994418 AK009997.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994418 AK010016.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994418 AK009566.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994418 AK009632.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P298259 AK009566.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120717 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30590759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12907678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12554365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30590750 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13023274 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10438568 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438569 - -
          Affymetrix HT MG-430 PM Array Plate 1423494_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10438567 - -
          Mouse Genome 430 2.0 Array 1423494_at - -
          Mouse Genome 430 2.0 Array 1423494_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66561_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66561_at - -
          Mouse Expression Set 430 1423494_PM_at - -
          Mouse Expression Set 430 1423494_at - -
          Murine Genome U74v2 Set 137017_PM_at - -
          Murine Genome U74v2 Set 137017_at - -
          Mouse Genome 430A 2.0 Array 1423494_at - -
          Mouse Genome 430A 2.0 Array 1423494_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423494_at - -
          Affymetrix HT Mouse Genome 430A Array 1423494_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5420707 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005420707 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2634140 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2634140 - -
          mouse-6 v1.1 (Illumina) scl066561.1_283-S - -
          Illumina Sentrix mouse-6.v1 bead array 5420707 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl066561.1_283-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5420707 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2634140 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2634140 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline