Search
Search

Asb13| Gene Symbol: Asb13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK008728.1 | - | 1 - 2 | 88 | 54 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
family with sequence similarity 208, member B |
Fam208b |
AI645998, Pap20, mKIAA2006 |
-22.9kb | Anti-sense | Chr. 13 - 3566035 - 3611231 on NCBI Build GRCm38 |
| Downstream | calmodulin-like 3 | Calml3 |
2310068O22Rik, AI326174 |
168.1kb | Anti-sense | Chr. 13 - 3802893 - 3804318 on NCBI Build GRCm38 |
Set Membership: |
|