Search
Search

Gemin4| Gene Symbol: Gemin4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK028076.1 | - | 1 - 2 | 39 | 70 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
family with sequence similarity 57, member A |
Fam57a |
2310047D13Rik, 4932415L08Rik, 5430402E13Rik, 5430420K21Rik, Wdt3 |
-7.7kb | Anti-sense | Chr. 11 - 76179676 - 76209417 on NCBI Build GRCm38 |
| Downstream | diazepam binding inhibitor-like 5 | Dbil5 | ELP | 98.0 | Anti-sense | Chr. 11 - 76217613 - 76218665 on NCBI Build GRCm38 |
Set Membership: |
|