Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03931958_m1
          See other 4933400F21RIK GE Assays ›
          Gene Symbol:
          4933400F21Rik
          Gene Name:
          RIKEN cDNA 4933400F21 gene
          Gene Aliases:
          1700091C04Rik, 4930565K10Rik
          Chromosome Location:
          Chr.1: 89676507 - 89683892 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03931958_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933400F21Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015540.1 1 - 2 201 100
          GenBank mRNA AK007030.1 - 1 - 2 198 100
          AK016229.1 - 220 100
          AK016592.1 - 1 - 2 203 100
          AK029704.1 - 1 - 2 205 101
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY715838.1 - 1 - 2 217 Chr. 1 - 89650641 - 89683908 [-] on Build GRCm38 100
          GenBankmRNA BB616064.1 - 1 - 2 181 Chr. 1 - 89680815 - 89683870 [-] on Build GRCm38 100
          GenBankmRNA CJ047535.1 - 1 - 2 207 Chr. 1 - 89681033 - 89683898 [-] on Build GRCm38 100
          GenBankmRNA BB615358.1 - 1 - 2 201 Chr. 1 - 89679411 - 89683892 [-] on Build GRCm38 100
          GenBankmRNA BB616490.1 - 1 - 2 193 Chr. 1 - 89680820 - 89683883 [-] on Build GRCm38 100

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA C030007H22 gene C030007H22Rik -275.0kb Sense Chr. 1 - 89360342 - 89406133 on NCBI Build GRCm38
          Downstream gastrulation brain homeobox 2 Gbx2 D130058E05Rik,
          Gbx-2,
          MMoxA,
          Stra7
          244.1kb Sense Chr. 1 - 89927962 - 89931176 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002780.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100022491.mm.1 NR_015540.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100022491.mm.1 NR_015540.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100022491.mm.1 TC0100002780.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100022490.mm.1 NR_015540.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100022490.mm.1 NR_015540.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100022490.mm.1 TC0100002780.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline