| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1445296_PM_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1436964_PM_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1455087_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1455087_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1455087_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1436964_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1436964_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1445296_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1445296_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0700028679.mm.1 |
NR_015456.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028679.mm.1 |
NR_015456.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028679.mm.1 |
TC0700003345.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700028677.mm.1 |
NR_015456.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028677.mm.1 |
NR_015456.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028677.mm.1 |
TC0700003345.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700028267.mm.1 |
NR_015479.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028267.mm.1 |
NR_015479.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028267.mm.1 |
TC0700003126.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700028270.mm.1 |
NR_015479.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028270.mm.1 |
NR_015479.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028270.mm.1 |
TC0700003126.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700028666.mm.1 |
NR_015456.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028666.mm.1 |
NR_015456.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028666.mm.1 |
TC0700003345.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700028271.mm.1 |
NR_015479.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028271.mm.1 |
NR_015479.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028271.mm.1 |
TC0700003126.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700028674.mm.1 |
NR_015456.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028674.mm.1 |
NR_015456.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028674.mm.1 |
TC0700003345.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700028667.mm.1 |
NR_015456.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028667.mm.1 |
NR_015456.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028667.mm.1 |
TC0700003345.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700028670.mm.1 |
NR_015456.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028670.mm.1 |
NR_015456.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028670.mm.1 |
TC0700003345.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700028675.mm.1 |
NR_015456.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028675.mm.1 |
NR_015456.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028675.mm.1 |
TC0700003345.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700028676.mm.1 |
NR_015456.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028676.mm.1 |
NR_015456.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028676.mm.1 |
TC0700003345.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700028680.mm.1 |
NR_015456.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028680.mm.1 |
NR_015456.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028680.mm.1 |
TC0700003345.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700028671.mm.1 |
NR_015456.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028671.mm.1 |
NR_015456.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028671.mm.1 |
TC0700003345.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700028668.mm.1 |
NR_015456.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028668.mm.1 |
NR_015456.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028668.mm.1 |
TC0700003345.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700028683.mm.1 |
NR_015456.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028683.mm.1 |
NR_015456.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028683.mm.1 |
TC0700003345.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700028264.mm.1 |
NR_015479.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028264.mm.1 |
NR_015479.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028264.mm.1 |
TC0700003126.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700028265.mm.1 |
NR_015479.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028265.mm.1 |
NR_015479.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028265.mm.1 |
TC0700003126.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700028669.mm.1 |
NR_015456.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028669.mm.1 |
NR_015456.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028669.mm.1 |
TC0700003345.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700028682.mm.1 |
NR_015456.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028682.mm.1 |
NR_015456.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028682.mm.1 |
TC0700003345.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1445296_PM_at |
- |
- |
| Mouse Expression Set 430 |
1436964_at |
- |
- |
| Mouse Expression Set 430 |
1455087_at |
- |
- |
| Mouse Expression Set 430 |
1436964_PM_at |
- |
- |
| Mouse Expression Set 430 |
1455087_PM_at |
- |
- |
| Mouse Expression Set 430 |
1445296_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1445296_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1436964_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1445296_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1436964_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1455087_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1455087_at |
- |
- |