Search
Search

Rapgef5| Gene Symbol: Rapgef5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK049664.1 | - | 1 | 2052 | 83 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 39559 |
Gm39559 | -392.6kb | Sense | Chr. 12 - 117345562 - 117347995 on NCBI Build GRCm38 | |
| Downstream | cell division cycle associated 7 like | Cdca7l |
BC006933, JPO2 |
100.1kb | Sense | Chr. 12 - 117843475 - 117878712 on NCBI Build GRCm38 |
Set Membership: |
|