Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03941001_s1
          See other 5430440L12RIK GE Assays ›
          Gene Symbol:
          5430440L12Rik
          Gene Name:
          RIKEN cDNA 5430440L12 gene
          Gene Aliases:
          AW536637
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03941001_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 5430440L12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AI157990.1 - 1 106
          AK017417.1 - 1 1152 106
          AW536637.1 - 1 106
          BG065027.2 - 1 106
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BE335859.1 - 1 Chr. 19 - 4794720 - 4795167 [+] on Build GRCm38 106
          GenBankmRNA BE947995.1 - 1 Chr. 19 - 4794715 - 4795280 [+] on Build GRCm38 106
          GenBankmRNA BB338102.1 - 1 199 Chr. 19 - 4794728 - 4795003 [-] on Build GRCm38 106
          GenBankmRNA CR519971.1 - 1 Chr. 19 - 4794717 - 4794941 [+] on Build GRCm38 106
          GenBankmRNA AV478375.1 - 1 358 Chr. 19 - 4794739 - 4795162 [-] on Build GRCm38 106
          GenBankmRNA AI480636.1 - 1 Chr. 19 - 4794716 - 4795189 [+] on Build GRCm38 106
          GenBankmRNA CF585815.1 - Chr. - - [-] on Build 106
          GenBankmRNA BM195683.2 - 1 Chr. 19 - 4794720 - 4794971 [+] on Build GRCm38 106
          GenBankmRNA AI158724.1 - 1 Chr. 19 - 4794715 - 4795082 [+] on Build GRCm38 106

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 21992 Gm21992 Rbm14-rbm4 Sense Chr. 19 - 4784293 - 4811634 on NCBI Build GRCm38
          Downstream RNA binding motif protein 14 Rbm14 1300007E16Rik,
          PSP2,
          Sytip,
          p16,
          p16K
          4.6kb Sense Chr. 19 - 4800566 - 4811634 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2380627 - -
          Whole Mouse Genome G4122A A_52_P962404 BG065027.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P962404 AI157990.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P962404 AK017417.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P962404 AW536637.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1433174_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1433173_PM_at - -
          Mouse Genome 430 2.0 Array 1433174_PM_a_at - -
          Mouse Genome 430 2.0 Array 1433173_PM_at - -
          Mouse Genome 430 2.0 Array 1433173_at - -
          Mouse Genome 430 2.0 Array 1433174_a_at - -
          Mouse Expression Set 430 1433174_a_at - -
          Mouse Expression Set 430 1433174_PM_a_at - -
          Mouse Expression Set 430 1433173_at - -
          Mouse Expression Set 430 1433173_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433174_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1433173_at - -
          Affymetrix HT Mouse Genome 430B Array 1433174_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1433173_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 692932 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline