Search
Search

Arhgap29| Gene Symbol: Arhgap29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK078431.1 | - | 1252 | 78 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ATP-binding cassette, sub-family D (ALD), member 3 |
Abcd3 |
AI313901, AU018866, AW146054, PMP68, PMP70, Pxmp1 |
-200.8kb | Anti-sense | Chr. 3 - 121758907 - 121815215 on NCBI Build GRCm38 |
| Downstream |
ATP-binding cassette, sub-family A (ABC1), member 4 |
Abca4 |
AW050280, Abc10, Abcr, D430003I15Rik, RmP |
27.1kb | Sense | Chr. 3 - 122043893 - 122180070 on NCBI Build GRCm38 |
Set Membership: |
|