Search
Search

4930578E11Rik| Gene Symbol: 4930578E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045391.1 | 5 - 6 | 1356 | 101 | ||
| GenBank mRNA | AK132929.1 | - | 5 - 6 | 1357 | 101 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4930527G23 gene | 4930527G23Rik | Anti-sense | Chr. 18 - 29442200 - 29466107 on NCBI Build GRCm38 | ||
| Downstream |
phosphoinositide-3-kinase, class 3 |
Pik3c3 |
5330434F23Rik, Vps34 |
828.1kb | Anti-sense | Chr. 18 - 30272094 - 30348126 on NCBI Build GRCm38 |
Set Membership: |
|