Search
Search

3110027N22Rik| Gene Symbol: 3110027N22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014088.1 | - | 1 | 649 | 85 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glycosyltransferase-like domain containing 1 | Gtdc1 | E330008O22Rik | -64.1kb | Sense | Chr. 2 - 44564412 - 44927918 on NCBI Build GRCm38 |
| Downstream | microRNA 5129 | Mir5129 |
mir-5129, mmu-mir-5129, mu-mir-5129 |
29.8kb | Sense | Chr. 2 - 45023100 - 45023177 on NCBI Build GRCm38 |
Set Membership: |
|