Search
Search
| Gene Symbol: Dep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF032130.1 | - | 1 | 2250 | 89 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | quaking | Qk |
1110003F05Rik, 1500005P18, QkI, l(17)-1Wis, l17Wis1 |
Sense | Chr. 17 - 10202856 - 10319361 on NCBI Build GRCm38 | |
| Downstream | colon adenocarcinoma hypermethylated RNA | Cahm | B930003M22Rik | 2.1kb | Anti-sense | Chr. 17 - 10319919 - 10321141 on NCBI Build GRCm38 |
Set Membership: |
|