Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03990486_g1
          See other TGM7 GE Assays ›
          Gene Symbol:
          Tgm7
          Gene Name:
          transglutaminase 7
          Gene Aliases:
          TGMZ, TGz
          Chromosome Location:
          Chr.2: 121093498 - 121127516 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03990486_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tgm7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001160424.1 NP_001153896.1 11 - 12 1976 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004478.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1997051 NM_001160424.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 66627 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 83871 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 30699 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 799627 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 515089 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 380066 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132317 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 835480 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 895064 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 380134 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 220949 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142400 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 451 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1997051 NM_001160424.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10486700 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486707 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486703 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486709 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486699 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486702 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486706 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486698 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486704 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486708 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486705 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486701 - -
          Affymetrix Mouse Gene 1.1 ST Array 10486697 - -
          Affymetrix Mouse Clariom S Array PSR0200037807.mm.1 NM_001160424.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200037807.mm.1 NM_001160424.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200037807.mm.1 TC0200004478.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225830 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline