Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03990708_m1
          See other CERS3 GE Assays ›
          Gene Symbol:
          Cers3
          Gene Name:
          ceramide synthase 3
          Gene Aliases:
          4930550L11Rik, Gm488, Lass3, T3L
          Chromosome Location:
          Chr.7: 66733178 - 66823697 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03990708_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cers3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164201.1 NP_001157673.1 10 - 11 1246 92
          GenBank mRNA DQ358087.1 - 9 - 10 1004 92
          DQ646881.1 - 10 - 11 1125 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000999.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          sphingolipid biosynthetic process
          keratinocyte differentiation
          ceramide biosynthetic process
          DNA binding
          transferase activity
          sphingosine N-acyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2325663 NM_001164201.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2325663 AV270649.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2325663 AI429073.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2099940 NM_001164201.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2099940 DQ358087.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2099940 DQ646881.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 836753 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 877536 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 326570 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777344 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 648433 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 472592 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 58264 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 68909 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 433131 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 876993 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 47864 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 940644 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 29283 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 697230 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 302729 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930005 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 516361 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 374584 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 134838 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 473807 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 573414 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 486853 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 285920 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 123446 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 574190 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 941978 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 454684 AK079397.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 454684 AK081206.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 454684 AK169810.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 454684 BC012512.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 454684 NM_175411.4 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 651146 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 174276 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 571356 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 340302 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674597 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 158339 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 681591 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 438789 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 282081 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 919734 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465534 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 411369 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 469180 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 402837 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 174281 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 902751 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 9372 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 780798 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 811112 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 894079 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 21375 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 686575 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 807313 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 258632 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 7957 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 798731 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 339368 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142762 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 4668 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 147000 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 103545 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 59755 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 170032 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 80861 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 102140 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35293 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2099940 NM_001164201.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2099940 DQ358087.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2099940 DQ646881.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2325663 NM_001164201.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2325663 AV270649.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2325663 AI429073.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06331302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06057648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06420703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06642973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25189960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06019469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06078952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06334989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25189959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06057667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25189991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06642978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25189970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05949514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25189995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06420702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25189990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25189967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25189962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06622158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06622157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25189982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25189981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05936474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25189978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25189988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25189999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25189983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25189985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25189987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06078961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25189973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25189972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25189964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25189977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06420700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05985015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06078949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25189971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06331296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06331286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25189969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06573272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25190349 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10554040 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554037 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554035 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554036 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554039 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554038 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554041 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554044 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554042 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554043 - -
          Affymetrix HT MG-430 PM Array Plate 1444740_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10554034 - -
          Affymetrix Mouse Clariom S Array PSR0700009181.mm.1 NM_001164201.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700009181.mm.1 NM_001164201.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700009181.mm.1 TC0700000999.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700009182.mm.1 NM_001164201.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700009182.mm.1 NM_001164201.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700009182.mm.1 TC0700000999.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1243498 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline