Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03990915_g1
          See other TSHB GE Assays ›
          Gene Symbol:
          Tshb
          Gene Name:
          thyroid stimulating hormone, beta subunit
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 102773624 - 102803490 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03990915_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tshb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001165939.1 NP_001159411.1 3 - 4 274 99
          NM_001165940.1 NP_001159412.1 2 - 3 227 99
          NM_009432.2 NP_033458.2 4 - 5 315 99
          GenBank mRNA BC116829.1 - 3 - 4 236 40126148 99
          BC116831.1 - 3 - 4 236 40126150 99
          BC144732.1 - 2 - 3 194 9053280 99
          J00644.1 - 3 - 4 248 99
          M54943.1 - 3 - 4 248 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002617.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tshb

          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Process(es)

          hormone activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P146711 BC144732.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P146711 J00644.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P146711 M54943.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P146711 NM_001165939.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P146711 NM_001165940.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P146711 BC144732.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P146711 J00644.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P146711 M54943.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P146711 NM_001165939.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P146711 NM_001165940.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 801722 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 278236 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 826465 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 147546 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142972 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P146711 BC144732.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P146711 J00644.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P146711 M54943.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P146711 NM_001165939.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P146711 NM_001165940.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P146711 BC144732.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P146711 J00644.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P146711 M54943.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P146711 NM_001165939.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P146711 NM_001165940.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P146711 BC144732.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P146711 J00644.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P146711 M54943.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P146711 NM_001165939.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P146711 NM_001165940.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P146711 BC144732.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P146711 J00644.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P146711 M54943.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P146711 NM_001165939.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P146711 NM_001165940.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22383024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22383033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22383014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22383019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22383016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22382997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22383021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22383009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22382995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22383002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22382996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22383022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22383028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22383013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22383006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22382994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22383027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22383032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22383037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22383029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22382999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22383011 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05012123 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142972 - -
          Whole Mouse Genome G4122A A_52_P146711 BC144732.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P146711 J00644.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P146711 M54943.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P146711 NM_001165939.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P146711 NM_001165940.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P146711 BC144732.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P146711 J00644.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P146711 M54943.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P146711 NM_001165939.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P146711 NM_001165940.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P252243 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140610 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142972 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02393204 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02393207 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03021704 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02393215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22382971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22383054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22382968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02755606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02749924 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02330153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02749921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02936361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22382985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02330150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22382956 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22382957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383018 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03177426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22382995 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22382983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02941292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22382975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02749916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22382953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22382988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22382996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22382979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22382974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02330163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02996974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02749919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22382951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02330147 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22382963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02453964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22382982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22382971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22382987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03021707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383013 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22382981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22382992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383035 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22382994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03106351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22382961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22383016 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10500750 - -
          Affymetrix Mouse Gene 1.0 ST Array 10500751 - -
          Affymetrix Mouse Gene 1.0 ST Array 10500749 - -
          Affymetrix Mouse Gene 1.0 ST Array 10500753 - -
          Affymetrix Mouse Gene 1.0 ST Array 10500752 - -
          Affymetrix HT MG-430 PM Array Plate 1450371_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10500748 - -
          Mouse Genome 430 2.0 Array 1450371_at - -
          Mouse Genome 430 2.0 Array 1450371_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22094_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22094_at - -
          Affymetrix Mouse Clariom S Array PSR0300020442.mm.1 NM_001165939.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020442.mm.1 NM_001165939.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020442.mm.1 NM_001165940.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020442.mm.1 NM_001165940.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020442.mm.1 NM_009432.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020442.mm.1 NM_009432.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020442.mm.1 TC0300002617.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300020444.mm.1 NM_001165939.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020444.mm.1 NM_001165939.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020444.mm.1 NM_001165940.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020444.mm.1 NM_001165940.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020444.mm.1 NM_009432.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020444.mm.1 NM_009432.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020444.mm.1 TC0300002617.mm.1 MTA TC
          Mouse Expression Set 430 1450371_PM_at - -
          Mouse Expression Set 430 1450371_at - -
          Murine Genome U74v2 Set 97741_PM_at - -
          Murine Genome U74v2 Set 97741_at - -
          Mouse Genome 430A 2.0 Array 1450371_at - -
          Mouse Genome 430A 2.0 Array 1450371_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450371_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450371_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 878281 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6770500 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006770500 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2464237 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2464237 - -
          mouse-6 v1.1 (Illumina) scl21745.5.1_164-S - -
          Illumina Sentrix mouse-6.v1 bead array 6770500 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21745.5.1_164-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6770500 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1215013 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215013 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline