| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100022381.mm.1 |
NM_198652.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022381.mm.1 |
NM_198652.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022381.mm.1 |
TC0100002772.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100022382.mm.1 |
NM_198652.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022382.mm.1 |
NM_198652.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022382.mm.1 |
TC0100002772.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100022385.mm.1 |
NM_198652.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022385.mm.1 |
NM_198652.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100022385.mm.1 |
TC0100002772.mm.1 |
MTA TC |