Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04176182_uH
          See other KLRA9 GE Assays ›
          Gene Symbol:
          Klra9
          Gene Name:
          killer cell lectin-like receptor subfamily A, member 9
          Gene Aliases:
          LY49I1, Ly49I
          Chromosome Location:
          Chr.6: 130178626 - 130194374 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04176182_uH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klra9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010651.3 NP_034781.2 1 114 160

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the NM_010651.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Inventoried Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klra9

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032718 U49867.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032718 AY860970.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032718 U34891.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032718 U34892.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2032718 U49865.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032718 U49867.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032718 AY860970.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032718 U34891.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032718 U34892.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2032718 U49865.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24846994 - -
          Whole Mouse Genome G4122A A_52_P361471 U34892.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P361471 AF237686.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05654472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05476451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846936 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10548533 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548515 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548507 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548518 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548526 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548524 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548516 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548506 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548522 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548534 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548527 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548519 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548521 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548514 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548517 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548529 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548528 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548530 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548523 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548531 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548520 - -
          Affymetrix Mouse Gene 1.1 ST Array 10548513 - -
          Affymetrix Mouse Gene 1.1 ST Array 10548525 - -
          Affymetrix Mouse Gene 1.1 ST Array 10548504 - -
          Affymetrix Mouse Gene 1.1 ST Array 10548532 - -
          Mouse Genome 430 2.0 Array 1425436_x_at - -
          Mouse Genome 430 2.0 Array 1425436_PM_x_at - -
          Affymetrix Murine Genome 6500A Array Msa16630_l_at - -
          Affymetrix Mouse Clariom S Array PSR0600025591.mm.1 NM_010651.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025591.mm.1 NM_010651.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025591.mm.1 TC0600003611.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025587.mm.1 NM_010651.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025587.mm.1 NM_010651.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025587.mm.1 TC0600003611.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025588.mm.1 NM_010651.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025588.mm.1 NM_010651.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025588.mm.1 TC0600003611.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025594.mm.1 NM_010651.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025594.mm.1 NM_010651.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025594.mm.1 TC0600003611.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025583.mm.1 NM_010651.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025583.mm.1 NM_010651.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025583.mm.1 TC0600003611.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025584.mm.1 NM_010651.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025584.mm.1 NM_010651.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025584.mm.1 TC0600003611.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025590.mm.1 NM_010651.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025590.mm.1 NM_010651.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025590.mm.1 TC0600003611.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025589.mm.1 NM_010651.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025589.mm.1 NM_010651.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025589.mm.1 TC0600003611.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025592.mm.1 NM_010651.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025592.mm.1 NM_010651.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025592.mm.1 TC0600003611.mm.1 MTA TC
          Mouse Expression Set 430 1425436_PM_x_at - -
          Mouse Expression Set 430 1425436_x_at - -
          Murine Genome U74v2 Set 93894_PM_f_at - -
          Murine Genome U74v2 Set 93894_f_at - -
          Murine Genome U74v2 Set 94779_f_at - -
          Murine Genome U74v2 Set 94779_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1425436_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1425436_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 858328 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101500161 - -
          MouseWG-6 v2 Expression BeadChip 730088 - -
          Illumina mouse-6 v1.0 expression beadchip 730088 - -
          Illumina mouse-6 v1.0 expression beadchip 101500161 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000730088 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2716458 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2613073 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2716458 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2613073 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233017 - -
          mouse-6 v1.1 (Illumina) scl016640.1_7-S - -
          mouse-6 v1.1 (Illumina) GI_31982782-S - -
          Illumina Sentrix mouse-6.v1 bead array 730088 - -
          Illumina Sentrix mouse-6.v1 bead array 101500161 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl016640.1_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 730088 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101500161 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1233017 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2613073 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline