Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04178105_g1
          See other OLFR1386 GE Assays ›
          Gene Symbol:
          Olfr1386
          Gene Name:
          olfactory receptor 1386
          Gene Aliases:
          MOR256-50
          Chromosome Location:
          Chr.11: 49470054 - 49471181 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04178105_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1386
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011741.2 NP_001011741.1 1 1056 92
          GenBank mRNA BC147372.1 - 1 1056 9056436 92
          BC147373.1 - 1 1056 9056431 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000512.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1386

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P91019 NM_001011741.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P91019 BC147372.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P91019 BC147373.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P91019 NM_001011741.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P91019 BC147372.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P91019 BC147373.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P91019 NM_001011741.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P91019 BC147372.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P91019 BC147373.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P91019 NM_001011741.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P91019 BC147372.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P91019 BC147373.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P91019 NM_001011741.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P91019 BC147372.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P91019 BC147373.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P91019 NM_001011741.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P91019 BC147372.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P91019 BC147373.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27783102 - -
          Whole Mouse Genome G4122A A_52_P91019 NM_001011741.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P91019 BC147372.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P91019 BC147373.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P91019 NM_001011741.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P91019 BC147372.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P91019 BC147373.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09207638 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09207585 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09207589 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09207586 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09207633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27783054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09207633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09268920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27783138 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10375564 - -
          Affymetrix Mouse Gene 1.0 ST Array 10375566 - -
          Affymetrix Mouse Gene 1.1 ST Array 10375565 - -
          Affymetrix Mouse Gene 1.1 ST Array 10375563 - -
          Affymetrix Mouse Clariom S Array PSR1100003907.mm.1 NM_001011741.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100003907.mm.1 NM_001011741.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100003907.mm.1 TC1100000512.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 540927 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2962701 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2962701 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline