Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04204434_mH
          See other GM20736 GE Assays ›
          Gene Symbol:
          Gm20736 1700040F15Rik Gm20857 Gm20858 Gm20871 Sly
          Gene Name:
          predicted gene, 20736
          RIKEN cDNA 1700040F15 gene
          Sycp3 like Y-linked pseudogene
          Sycp3 like Y-linked pseudogene
          Sycp3 like Y-linked pseudogene
          Sycp3 like Y-linked
          Gene Aliases:
          -
          -
          -
          -
          -
          -
          Chromosome Location:
          Chr.Y: 65377690 - 65403913 on Build GRCm38
          Chromosome Location:
          Chr.Y: 60241398 - 60297384 on Build GRCm38
          Chromosome Location:
          Chr.Y: 25214873 - 25240906 on Build GRCm38
          Chromosome Location:
          Chr.Y: 25727935 - 25753940 on Build GRCm38
          Chromosome Location:
          Chr.Y: 59606710 - 59632758 on Build GRCm38
          Chromosome Location:
          Chr.Y: 57187206 - 57213358 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          Assay ID Mm04204434_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm20736
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037748.1 NP_001032837.2 3 - 4 257 105
          GenBank mRNA BC100464.1 - 3 - 4 257 6705131 105
          Gene Symbol: 1700040F15Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA FJ541086.1 - 171 238
          Gene Symbol: Gm20857
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038297.1 257 140
          GenBank mRNA BU945722.1 - 260 140
          Gene Symbol: Gm20858
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038298.1 257 140
          Gene Symbol: Gm20871
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038299.1 3 - 4 232 105
          GenBank mRNA BC099537.1 - 232 30916067 105
          Gene Symbol: Sly
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_201530.2 NP_963288.2 3 - 4 242 207
          GenBank mRNA BC049626.1 - 3 - 4 242 6705850 207

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0Y00000091.mm.1, TC0Y00000154.mm.1, TC0Y00000206.mm.1, TC0Y00000283.mm.1, TC0Y00000284.mm.1, TC0Y00000285.mm.1, TC0Y00000288.mm.1, TC0Y00000558.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gm20736
          Click here to find siRNAs for Gene 1700040F15Rik
          Click here to find siRNAs for Gene Gm20857
          Click here to find siRNAs for Gene Gm20858
          Click here to find siRNAs for Gene Gm20871
          Click here to find siRNAs for Gene Sly

          Gene Ontology Categories:

          Function(s) Process(es)

          spermatid development
          single fertilization
          sex determination
          regulation of gene expression
          spermatid differentiation
          protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1975546 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1975546 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10608592 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608493 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608593 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608256 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608587 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608496 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608525 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608497 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608621 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608445 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608398 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608253 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608618 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608529 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608624 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608595 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608246 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608471 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608444 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608473 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608357 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608243 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608258 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608588 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608240 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608467 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608433 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608245 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608331 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608468 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608253 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608333 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608405 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608623 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608622 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608403 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608334 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608435 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608589 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608239 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608617 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608619 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608498 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608447 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608238 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608241 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608358 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608495 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608446 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608399 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608259 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608472 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608244 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608465 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608360 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608252 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608432 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608257 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608406 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608256 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608437 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608431 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608259 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608258 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608356 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608527 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608335 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608354 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608400 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608590 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608336 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608436 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608404 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608528 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608466 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608526 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608254 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608530 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608434 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608616 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608359 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608494 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608355 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608337 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608254 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608594 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608401 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608430 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608257 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608252 - -
          Affymetrix Mouse Gene 1.1 ST Array 10608586 - -
          Affymetrix Mouse Gene 1.1 ST Array 10608492 - -
          Affymetrix Mouse Gene 1.1 ST Array 10608442 - -
          Affymetrix Mouse Gene 1.1 ST Array 10608429 - -
          Affymetrix Mouse Gene 1.1 ST Array 10608251 - -
          Affymetrix Mouse Gene 1.1 ST Array 10608523 - -
          Affymetrix Mouse Clariom S Array PSR0Y00000545.mm.1 NR_038299.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000545.mm.1 TC0Y00000558.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000715.mm.1 NM_201530.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000715.mm.1 TC0Y00000154.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000721.mm.1 NM_201530.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000721.mm.1 TC0Y00000154.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001102.mm.1 NM_001037748.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001102.mm.1 TC0Y00000206.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001651.mm.1 NR_038297.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001651.mm.1 TC0Y00000285.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001666.mm.1 NR_038298.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001666.mm.1 TC0Y00000288.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001672.mm.1 NR_038298.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001672.mm.1 TC0Y00000288.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000394.mm.1 TC0Y00000091.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000547.mm.1 NR_038299.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000547.mm.1 TC0Y00000558.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000554.mm.1 NR_038299.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000554.mm.1 TC0Y00000558.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000557.mm.1 NR_038299.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000557.mm.1 TC0Y00000558.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000718.mm.1 NM_201530.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000718.mm.1 TC0Y00000154.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001645.mm.1 NR_038297.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001645.mm.1 TC0Y00000285.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000391.mm.1 TC0Y00000091.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001637.mm.1 TC0Y00000284.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000714.mm.1 NM_201530.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000714.mm.1 TC0Y00000154.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001104.mm.1 NM_001037748.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001104.mm.1 TC0Y00000206.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000398.mm.1 TC0Y00000091.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001627.mm.1 TC0Y00000283.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001633.mm.1 TC0Y00000284.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000717.mm.1 NM_201530.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000717.mm.1 TC0Y00000154.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001106.mm.1 NM_001037748.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001106.mm.1 TC0Y00000206.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001647.mm.1 NR_038297.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001647.mm.1 TC0Y00000285.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001654.mm.1 NR_038297.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001654.mm.1 TC0Y00000285.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001624.mm.1 TC0Y00000283.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001631.mm.1 TC0Y00000284.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001634.mm.1 TC0Y00000284.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000546.mm.1 NR_038299.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000546.mm.1 TC0Y00000558.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000555.mm.1 NR_038299.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000555.mm.1 TC0Y00000558.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000720.mm.1 NM_201530.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000720.mm.1 TC0Y00000154.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001105.mm.1 NM_001037748.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001105.mm.1 TC0Y00000206.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001649.mm.1 NR_038297.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001649.mm.1 TC0Y00000285.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001653.mm.1 NR_038297.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001653.mm.1 TC0Y00000285.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001668.mm.1 NR_038298.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001668.mm.1 TC0Y00000288.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000390.mm.1 TC0Y00000091.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000393.mm.1 TC0Y00000091.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000399.mm.1 TC0Y00000091.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001623.mm.1 TC0Y00000283.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000543.mm.1 NR_038299.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000543.mm.1 TC0Y00000558.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000716.mm.1 NM_201530.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000716.mm.1 TC0Y00000154.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001652.mm.1 NR_038297.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001652.mm.1 TC0Y00000285.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001674.mm.1 NR_038298.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001674.mm.1 TC0Y00000288.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001675.mm.1 NR_038298.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001675.mm.1 TC0Y00000288.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000392.mm.1 TC0Y00000091.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000397.mm.1 TC0Y00000091.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001103.mm.1 NM_001037748.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001103.mm.1 TC0Y00000206.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001648.mm.1 NR_038297.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001648.mm.1 TC0Y00000285.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001669.mm.1 NR_038298.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001669.mm.1 TC0Y00000288.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001671.mm.1 NR_038298.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001671.mm.1 TC0Y00000288.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001625.mm.1 TC0Y00000283.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001626.mm.1 TC0Y00000283.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001632.mm.1 TC0Y00000284.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001636.mm.1 TC0Y00000284.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001638.mm.1 TC0Y00000284.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000550.mm.1 NR_038299.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000550.mm.1 TC0Y00000558.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000553.mm.1 NR_038299.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000553.mm.1 TC0Y00000558.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000556.mm.1 NR_038299.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000556.mm.1 TC0Y00000558.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00000719.mm.1 NM_201530.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00000719.mm.1 TC0Y00000154.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001099.mm.1 NM_001037748.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001099.mm.1 TC0Y00000206.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001650.mm.1 NR_038297.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001650.mm.1 TC0Y00000285.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001670.mm.1 NR_038298.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001670.mm.1 TC0Y00000288.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001673.mm.1 NR_038298.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001673.mm.1 TC0Y00000288.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001635.mm.1 TC0Y00000284.mm.1 MTA TC
          Murine Genome U74v2 Set 168238_PM_at - -
          Murine Genome U74v2 Set 168238_at - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2852764 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2852764 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline