Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04204903_gH
          See other GM13271 GE Assays ›
          Gene Symbol:
          Gm13271 Gm13274
          Gene Name:
          predicted gene 13271
          predicted gene 13274
          Gene Aliases:
          Ifnz, OTTMUSG00000011262
          OTTMUSG00000011267
          Chromosome Location:
          Chr.4: 88754868 - 88755416 on Build GRCm38
          Chromosome Location:
          Chr.4: 88723681 - 88744717 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm04204903_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm13271
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001085528.1 NP_001078997.1 1 56 92
          Gene Symbol: Gm13274
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK144774.1 - 92

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          interferon superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          T cell activation involved in immune response
          natural killer cell activation involved in immune response
          humoral immune response
          cytokine-mediated signaling pathway
          B cell differentiation
          positive regulation of peptidyl-serine phosphorylation of STAT protein
          B cell proliferation
          response to exogenous dsRNA
          cytokine activity
          type I interferon receptor binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2075613 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2141740 AK144774.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2141740 NM_001085528.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063402 AK144774.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063402 NM_001085532.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063402 NM_001098840.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063402 NM_001098841.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063402 NM_001085533.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2075613 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2141740 AK144774.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2141740 NM_001085528.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063402 AK144774.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063402 NM_001085532.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063402 NM_001098840.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063402 NM_001098841.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063402 NM_001085533.2 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23092005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03337854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03585858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091995 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03933739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03384752 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10505850 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505871 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505854 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505857 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505846 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505863 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505867 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505844 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505842 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505875 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505869 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505878 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505865 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505860 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505873 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505862 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505872 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505866 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505856 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505874 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505858 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505864 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505876 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505841 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505868 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505843 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505848 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505870 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505845 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505852 - -
          Affymetrix Mouse Clariom S Array PSR0400006046.mm.1 NM_001085528.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006046.mm.1 NM_001085528.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006046.mm.1 TC0400000889.mm.1 MTA TC
          Mouse Expression Set 430 1425322_PM_at - -
          Mouse Expression Set 430 1425322_at - -
          Mouse Genome 430A 2.0 Array 1425322_at - -
          Mouse Genome 430A 2.0 Array 1425322_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline