Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04205023_gH
          See other GM13278 GE Assays ›
          Gene Symbol:
          Gm13278 Gm13276 Gm13279 Gm13288 Gm13289 Gm13290 Ifnz
          Gene Name:
          predicted gene 13278
          predicted gene 13276
          predicted gene 13279
          predicted gene 13288
          predicted gene 13289
          predicted gene 13290
          interferon zeta
          Gene Aliases:
          OTTMUSG00000011272
          OTTMUSG00000011269
          OTTMUSG00000011273
          OTTMUSG00000011289
          OTTMUSG00000011290
          OTTMUSG00000011291
          6030405N23Rik, BGIF, ENSMUSG00000073810, Lmtn
          Chromosome Location:
          Chr.4: 88790895 - 88792356 on Build GRCm38
          Chromosome Location:
          Chr.4: 88785054 - 88786515 on Build GRCm38
          Chromosome Location:
          Chr.4: 88797237 - 88798204 on Build GRCm38
          Chromosome Location:
          Chr.4: 88805531 - 88808380 on Build GRCm38
          Chromosome Location:
          Chr.4: 88776762 - 88777729 on Build GRCm38
          Chromosome Location:
          Chr.4: 88773834 - 88774801 on Build GRCm38
          Chromosome Location:
          Chr.4: 88782131 - 88783592 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          Assay ID Mm04205023_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm13278
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001098841.3 NP_001092311.1 1 660 80
          Gene Symbol: Gm13276
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001085532.3 NP_001079001.1 1 660 80
          Gene Symbol: Gm13279
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001243166.1 NP_001230095.1 1 165 80
          Gene Symbol: Gm13288
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001243167.1 NP_001230096.1 1 660 80
          GenBank mRNA AK031317.1 - 1 661 80
          Gene Symbol: Gm13289
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001243165.1 NP_001230094.1 165 80
          Gene Symbol: Gm13290
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001243155.1 NP_001230084.1 1 165 80
          GenBank mRNA AB024521.1 - 1 165 80
          Gene Symbol: Ifnz
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_197889.2 NP_922871.1 1 660 80
          GenBank mRNA AK046550.1 - 1 661 80

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AB024521.1, AK031317.1, AK046550.1, NM_001085532.3, NM_001098841.3, NM_001243155.1, NM_001243165.1, NM_001243166.1, NM_001243167.1, NM_197889.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gm13278
          Click here to find siRNAs for Gene Gm13276
          Click here to find siRNAs for Gene Gm13279
          Click here to find siRNAs for Gene Gm13288
          Click here to find siRNAs for Gene Gm13289
          Click here to find siRNAs for Gene Gm13290
          Click here to find siRNAs for Gene Ifnz

          Panther Classification:

          Molecular Function -

          interferon superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          T cell activation involved in immune response
          natural killer cell activation involved in immune response
          humoral immune response
          biological_process
          cytokine-mediated signaling pathway
          B cell differentiation
          positive regulation of peptidyl-serine phosphorylation of STAT protein
          B cell proliferation
          response to exogenous dsRNA
          regulation of cell cycle
          apoptotic signaling pathway
          molecular_function
          cytokine activity
          type I interferon receptor binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline