Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04205241_mH
          See other KLRA8 GE Assays ›
          Gene Symbol:
          Klra8 Klra21 Klra3
          Gene Name:
          killer cell lectin-like receptor, subfamily A, member 8
          killer cell lectin-like receptor subfamily A, member 21
          killer cell lectin-like receptor, subfamily A, member 3
          Gene Aliases:
          Cmv-1, Cmv1, Ly49h, Ly49u<129>
          Klra-ps1, Ly49H, Ly49u
          5E6, Ly49c, NK-2.1, Nk-2, Nk2, Nk2.1
          Chromosome Location:
          Chr.6: 130115226 - 130129898 on Build GRCm38
          Chromosome Location:
          Chr.6: 130082598 - 130096486 on Build GRCm38
          Chromosome Location:
          Chr.6: 130323289 - 130337626 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Mm04205241_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klra8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001101620.1 NP_001095090.1 2 - 3 259 92
          GenBank mRNA U12889.1 - 2 - 3 310 92
          Gene Symbol: Klra21
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053151.1 NP_444381.1 226 92
          GenBank mRNA AF288380.1 - 226 92
          AY971806.1 - 120 92
          BC116400.1 - 204 40101979 92
          Gene Symbol: Klra3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC141217.1 - 210 9055829 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003610.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klra8
          Click here to find siRNAs for Gene Klra21
          Click here to find siRNAs for Gene Klra3

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          response to virus
          biological_process
          carbohydrate binding
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Whole Mouse Genome G4122A A_52_P680616 NM_053151.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P680616 AF288380.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P680616 BC116399.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05148344 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10548507 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548508 - -
          Affymetrix HT MG-430 PM Array Plate 1426136_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10548504 - -
          Mouse Genome 430 2.0 Array 1426136_PM_x_at - -
          Mouse Genome 430 2.0 Array 1426136_x_at - -
          Affymetrix Mouse Clariom S Array PSR0600025559.mm.1 NM_001101620.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025559.mm.1 NM_001101620.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025559.mm.1 NM_010650.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025559.mm.1 NM_010650.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025559.mm.1 TC0600003610.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025553.mm.1 TC0600003610.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025560.mm.1 NM_001101620.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025560.mm.1 NM_001101620.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025560.mm.1 NM_010650.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025560.mm.1 NM_010650.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025560.mm.1 TC0600003610.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025579.mm.1 NR_104105.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025579.mm.1 NR_104106.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025579.mm.1 TC0600003610.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025581.mm.1 TC0600003610.mm.1 MTA TC
          Mouse Expression Set 430 1426136_PM_x_at - -
          Mouse Expression Set 430 1426136_x_at - -
          Murine Genome U74v2 Set 93894_PM_f_at - -
          Murine Genome U74v2 Set 94779_PM_f_at - -
          Murine Genome U74v2 Set 94779_f_at - -
          Murine Genome U74v2 Set 93894_f_at - -
          Murine Genome U74v2 Set 134769_PM_f_at - -
          Murine Genome U74v2 Set 134769_f_at - -
          Mouse Genome 430A 2.0 Array 1426136_x_at - -
          Mouse Genome 430A 2.0 Array 1426136_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1426136_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1426136_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 326605 - -
          ABI Mouse Genome Survey Microarray 924342 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 7000671 - -
          MouseWG-6 v2 Expression BeadChip 5670402 - -
          Illumina mouse-6 v1.0 expression beadchip 105050348 - -
          Illumina mouse-6 v1.0 expression beadchip 7000671 - -
          Illumina mouse-6 v1.0 expression beadchip 5670402 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007000671 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005670402 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1236777 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219419 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1236777 - -
          mouse-6 v1.1 (Illumina) GI_21361215-S - -
          mouse-6 v1.1 (Illumina) scl0093968.1_91-S - -
          mouse-6 v1.1 (Illumina) scl093968.1_17-S - -
          Illumina Sentrix mouse-6.v1 bead array 7000671 - -
          Illumina Sentrix mouse-6.v1 bead array 105050348 - -
          Illumina Sentrix mouse-6.v1 bead array 5670402 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0093968.1_91-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl093968.1_17-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5670402 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7000671 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105050348 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1236777 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1252006 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1252006 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1219419 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1236777 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline