Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04205285_gH
          See other GM13242 GE Assays ›
          Gene Symbol:
          Gm13242 Rex2 Zfp600
          Gene Name:
          predicted gene 13242
          reduced expression 2
          zinc finger protein 600
          Gene Aliases:
          OTTMUSG00000011070
          Gm13138, OTTMUSG00000010655
          Gm13164, OTTMUSG00000010694
          Chromosome Location:
          Chr.4: 145670697 - 145704437 on Build GRCm38
          Chromosome Location:
          Chr.4: 147021850 - 147060799 on Build GRCm38
          Chromosome Location:
          Chr.4: 146156827 - 146198756 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Mm04205285_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm13242
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001103158.2 NP_001096628.2 3 - 4 330 92
          GenBank mRNA BC096593.1 - 3 - 4 342 30459933 92
          Gene Symbol: Rex2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177767.1 NP_001171238.1 3 - 4 330 92
          GenBank mRNA BC067397.1 - 1 - 2 85 30462971 92
          CD542867.1 - 3 - 4 314 92
          CF162675.1 - 3 - 4 314 92
          Gene Symbol: Zfp600
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177545.2 NP_001171016.1 3 - 4 237 92
          NM_001177546.2 NP_001171017.1 3 - 4 330 92
          GenBank mRNA CD539901.1 - 3 - 4 330 92

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC067397.1, BC096593.1 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          regulation of transcription, DNA-templated
          transcription factor activity, sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2037072 NM_001177545.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2037072 NM_001177546.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2037072 BC108352.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10510205 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510209 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510202 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510210 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510206 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510161 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510207 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510208 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510211 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510160 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510203 - -
          Affymetrix Mouse Gene 1.1 ST Array 10510201 - -
          Affymetrix Mouse Gene 1.1 ST Array 10510159 - -
          Affymetrix Mouse Clariom S Array PSR0400015585.mm.1 NM_001177767.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015585.mm.1 NM_001177767.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015585.mm.1 TC0400001894.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015521.mm.1 NM_001177545.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015521.mm.1 NM_001177546.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015521.mm.1 TC0400001878.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015482.mm.1 NM_001103158.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015482.mm.1 NM_001103158.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015482.mm.1 TC0400001869.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015488.mm.1 NM_001103158.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015488.mm.1 NM_001103158.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015488.mm.1 TC0400001869.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015513.mm.1 NM_001177546.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015513.mm.1 TC0400001878.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015522.mm.1 NM_001177545.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015522.mm.1 NM_001177546.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015522.mm.1 TC0400001878.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015579.mm.1 NM_001177767.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015579.mm.1 NM_001177767.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015579.mm.1 TC0400001894.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015514.mm.1 NM_001177545.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015514.mm.1 TC0400001878.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015515.mm.1 NM_001177545.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015515.mm.1 NM_001177546.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015515.mm.1 TC0400001878.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015583.mm.1 NM_001177767.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015583.mm.1 NM_001177767.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015583.mm.1 TC0400001894.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015520.mm.1 NM_001177545.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015520.mm.1 NM_001177546.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015520.mm.1 TC0400001878.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015580.mm.1 NM_001177767.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015580.mm.1 NM_001177767.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015580.mm.1 TC0400001894.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015485.mm.1 NM_001103158.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015485.mm.1 NM_001103158.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015485.mm.1 TC0400001869.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015519.mm.1 NM_001177545.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015519.mm.1 NM_001177546.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015519.mm.1 TC0400001878.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015581.mm.1 NM_001177767.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015581.mm.1 NM_001177767.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015581.mm.1 TC0400001894.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015483.mm.1 NM_001103158.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015483.mm.1 NM_001103158.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015483.mm.1 TC0400001869.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015486.mm.1 NM_001103158.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015486.mm.1 NM_001103158.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015486.mm.1 TC0400001869.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015487.mm.1 NM_001103158.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015487.mm.1 NM_001103158.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015487.mm.1 TC0400001869.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015584.mm.1 NM_001177767.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015584.mm.1 NM_001177767.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015584.mm.1 TC0400001894.mm.1 MTA TC
          Mouse Expression Set 430 1438237_at - -
          Mouse Expression Set 430 1438237_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline