Search
Search
| Gene Symbol: Asb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039126.2 | NP_001034215.1 | 3 - 4 | 748 | 104 | |
| NM_023046.4 | NP_075533.3 | 3 - 4 | 549 | 104 | ||
| GenBank mRNA | AF155352.1 | - | 3 - 4 | 500 | 104 | |
| AK006515.1 | - | 3 - 4 | 550 | 104 | ||
| AK122456.1 | - | 3 - 4 | 604 | 104 | ||
| AK153774.1 | - | 3 - 4 | 770 | 104 | ||
| BC034869.1 | - | 4 - 5 | 543 | 104 | ||
| BC049699.1 | - | 3 - 4 | 549 | 104 | ||
| BU938650.1 | - | 3 - 4 | 552 | 104 | ||
Set Membership: |
|