Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04206589_gH
          See other GM2933 GE Assays ›
          Gene Symbol:
          Gm2933 Gm10922 Gm14345 Gm14346 Gm14351 Gm2913 Gm2927 Gm3701 Gm3706 Gm3750 Gm3763
          Gene Name:
          predicted gene 2933
          predicted gene 10922
          predicted gene 14345
          predicted gene 14346
          predicted gene 14351
          predicted gene 2913
          predicted gene 2927
          predicted gene 3701
          predicted gene 3706
          predicted gene 3750
          predicted gene 3763
          Gene Aliases:
          -
          ENSMUSG00000078216, OTTMUSG00000016437
          OTTMUSG00000016405
          OTTMUSG00000016406
          OTTMUSG00000016411
          -
          -
          -
          -
          -
          -
          Chromosome Location:
          Chr.X: 33700649 - 33702349 on Build GRCm38
          Chromosome Location:
          Chr.X: 4370636 - 4372595 on Build GRCm38
          Chromosome Location:
          Chr.X: 3700233 - 3702192 on Build GRCm38
          Chromosome Location:
          Chr.X: 3441731 - 3443690 on Build GRCm38
          Chromosome Location:
          Chr.X: 3750931 - 3752885 on Build GRCm38
          Chromosome Location:
          Chr.X: 33575387 - 33577346 on Build GRCm38
          Chromosome Location:
          Chr.X: 33657140 - 33659082 on Build GRCm38
          Chromosome Location:
          Chr.X: 3955043 - 3957002 on Build GRCm38
          Chromosome Location:
          Chr.X: 4037674 - 4039622 on Build GRCm38
          Chromosome Location:
          Chr.X: 4800411 - 4802359 on Build GRCm38
          Chromosome Location:
          Chr.X: 4952135 - 4954077 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          Assay ID Mm04206589_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm2933
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145038.1 NP_001138510.1 1 354 103
          GenBank mRNA BC115962.1 - 1 354 40057634 103
          Gene Symbol: Gm10922
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001085554.1 NP_001079023.1 1 386 103
          Gene Symbol: Gm14345
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001085545.1 NP_001079014.1 1 386 103
          Gene Symbol: Gm14346
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001085551.1 NP_001079020.1 1 386 103
          Gene Symbol: Gm14351
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001085552.2 NP_001079021.1 1 386 103
          Gene Symbol: Gm2913
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001243015.1 NP_001229944.1 1 386 103
          Gene Symbol: Gm2927
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001282034.1 NP_001268963.1 1 386 103
          Gene Symbol: Gm3701
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001243011.1 NP_001229940.1 1 386 103
          Gene Symbol: Gm3706
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001243001.1 NP_001229930.1 1 386 103
          Gene Symbol: Gm3750
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001099643.2 NP_001093113.1 1 386 103
          Gene Symbol: Gm3763
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001243024.1 NP_001229953.1 1 386 103
          GenBank mRNA BC089532.1 - 1 386 6704091 103

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10603236 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603235 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598219 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603240 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598226 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603239 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603241 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603234 - -
          Affymetrix Mouse Gene 1.1 ST Array 10603232 - -
          Affymetrix Mouse Gene 1.1 ST Array 10603237 - -
          Affymetrix Mouse Gene 1.1 ST Array 10598218 - -
          Affymetrix Mouse Gene 1.1 ST Array 10598183 - -
          Affymetrix Mouse Clariom S Array PSR0X00000033.mm.1 NM_001085554.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00000033.mm.1 NM_001085554.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00000033.mm.1 TC0X00000021.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00001964.mm.1 NM_001282034.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001964.mm.1 TC0X00000296.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00001965.mm.1 NM_001282034.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001965.mm.1 TC0X00000296.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00012905.mm.1 NM_001145038.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00012905.mm.1 NM_001243015.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00012905.mm.1 TC0X00003421.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00000055.mm.1 NM_001243024.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00000055.mm.1 NM_001243024.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00000055.mm.1 TC0X00000032.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00000019.mm.1 NM_001243001.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00000019.mm.1 NM_001243001.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00000019.mm.1 TC0X00000012.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00011020.mm.1 NM_001085545.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011020.mm.1 NM_001085551.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011020.mm.1 NM_001085552.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011020.mm.1 NM_001099643.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011020.mm.1 NM_001243011.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011020.mm.1 TC0X00001723.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00011019.mm.1 NM_001085545.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011019.mm.1 NM_001085551.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011019.mm.1 NM_001085552.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011019.mm.1 NM_001099643.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011019.mm.1 NM_001243011.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011019.mm.1 TC0X00001723.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00011026.mm.1 NM_001085551.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011026.mm.1 NM_001243015.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011026.mm.1 TC0X00001725.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00011025.mm.1 NM_001085551.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011025.mm.1 NM_001243015.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011025.mm.1 TC0X00001725.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline