Search
Search

Ncald| Gene Symbol: Ncald | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170866.1 | NP_001164337.1 | 6 - 7 | 782 | 106 | |
| NM_001170867.1 | NP_001164338.1 | 4 - 5 | 716 | 106 | ||
| NM_001170868.1 | NP_001164339.1 | 3 - 4 | 664 | 106 | ||
| NM_134094.4 | NP_598855.2 | 6 - 7 | 972 | 106 | ||
| GenBank mRNA | AK034031.1 | - | 4 - 5 | 695 | 106 | |
| AK048440.1 | - | 6 - 7 | 952 | 106 | ||
| AK167313.1 | - | 3 - 4 | 634 | 106 | ||
| BC026979.1 | - | 3 - 4 | 618 | 5365560 | 106 | |
Set Membership: |
|