Search
Search

Ndufa4| Gene Symbol: Ndufa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010886.2 | NP_035016.1 | 2 - 3 | 210 | 93 | |
| GenBank mRNA | AK005084.1 | - | 2 - 3 | 215 | 93 | |
| AK008357.1 | - | 2 - 3 | 207 | 93 | ||
| AK008506.1 | - | 2 - 3 | 201 | 93 | ||
| AK076036.1 | - | 2 - 3 | 211 | 93 | ||
| BC011114.1 | - | 2 - 3 | 173 | 3483028 | 93 | |
| U59509.1 | - | 2 - 3 | 108 | 93 | ||
Set Membership: |
|