| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600004338.mm.1 |
NM_001243199.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004338.mm.1 |
NM_001243199.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004338.mm.1 |
NM_001243200.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004338.mm.1 |
NM_001243200.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004338.mm.1 |
NM_001243201.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004338.mm.1 |
NM_001243201.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004338.mm.1 |
NM_174990.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004338.mm.1 |
NM_174990.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004338.mm.1 |
NM_175048.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004338.mm.1 |
NM_175048.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004338.mm.1 |
TC0600000566.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600004339.mm.1 |
NM_001243199.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004339.mm.1 |
NM_001243199.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004339.mm.1 |
NM_001243200.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004339.mm.1 |
NM_001243200.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004339.mm.1 |
NM_001243201.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004339.mm.1 |
NM_001243201.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004339.mm.1 |
NM_174990.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004339.mm.1 |
NM_174990.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004339.mm.1 |
NM_175048.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004339.mm.1 |
NM_175048.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004339.mm.1 |
TC0600000566.mm.1 |
MTA TC |