Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04213467_gH
          See other ZNF41-PS GE Assays ›
          Gene Symbol:
          Znf41-ps
          Gene Name:
          ZNF41, pseudogene
          Gene Aliases:
          1700029I01Rik
          Chromosome Location:
          Chr.4: 145789515 - 145831978 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04213467_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Znf41-ps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040355.1 5 4351 99
          GenBank mRNA BC096621.1 - 4363 30464103 99

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P07455 NM_027285.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P07455 BC096621.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2121151 NM_027285.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2121151 BC096621.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P07455 NM_027285.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P07455 BC096621.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2121151 NM_027285.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2121151 BC096621.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109496 NM_001083918.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109496 BC024323.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109496 NM_001085522.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109496 NM_027285.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109496 BC096621.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P487156 NM_001127189.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P487156 BC055069.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1042412 NM_001205101.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P581100 NM_001083918.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P581100 BC024323.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P581100 AV583921.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P581100 NM_027285.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P581100 BC096621.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118350 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131400 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128170 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124735 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04033976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03949820 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10510168 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510182 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518338 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518337 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518336 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510169 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518348 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518347 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510183 - -
          Affymetrix Mouse Gene 1.0 ST Array 10490855 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518349 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510184 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510185 - -
          Affymetrix HT MG-430 PM Array Plate 1440518_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1436574_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10518346 - -
          Affymetrix Mouse Gene 1.1 ST Array 10518335 - -
          Mouse Genome 430 2.0 Array 1436574_PM_at - -
          Mouse Genome 430 2.0 Array 1436574_at - -
          Mouse Genome 430 2.0 Array 1424784_PM_at - -
          Mouse Genome 430 2.0 Array 1424784_at - -
          Mouse Genome 430 2.0 Array 1440518_at - -
          Mouse Genome 430 2.0 Array 1440518_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400015327.mm.1 NR_040355.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015327.mm.1 NR_040355.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015327.mm.1 TC0400004198.mm.1 MTA TC
          Mouse Expression Set 430 1445924_at - -
          Mouse Expression Set 430 1436574_PM_at - -
          Mouse Expression Set 430 1445924_PM_at - -
          Mouse Expression Set 430 1436574_at - -
          Murine Genome U74v2 Set 170148_PM_at - -
          Murine Genome U74v2 Set 170277_at - -
          Murine Genome U74v2 Set 170277_PM_at - -
          Murine Genome U74v2 Set 170148_at - -
          Affymetrix HT Mouse Genome 430B Array 1440518_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436574_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436574_at - -
          Affymetrix HT Mouse Genome 430B Array 1440518_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1990280 - -
          Illumina mouse-6 v1.0 expression beadchip 1990280 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001990280 - -
          mouse-6 v1.1 (Illumina) scl070005.1_316-S - -
          Illumina Sentrix mouse-6.v1 bead array 1990280 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl070005.1_316-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1990280 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2882698 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2592681 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2882698 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline