Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04214157_s1
          See other OLFR106-PS GE Assays ›
          Gene Symbol:
          Olfr106-ps
          Gene Name:
          olfactory receptor 106, pseudogene
          Gene Aliases:
          MOR250-6, Olfr106
          Chromosome Location:
          Chr.17: 37394590 - 37395524 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04214157_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr106-ps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC145568.1 - 1 570 102

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168529 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13517538 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13517551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13502108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13517548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13374392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13096090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168532 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445132 - -
          Affymetrix Mouse Gene 1.1 ST Array 10445131 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161068 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161068 - -
          mouse-6 v1.1 (Illumina) GI_58801311-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3161068 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3161068 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline