Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04243440_m1
          See other SBK3 GE Assays ›
          Gene Symbol:
          Sbk3
          Gene Name:
          SH3 domain binding kinase family, member 3
          Gene Aliases:
          Gm1078, Sgk110
          Chromosome Location:
          Chr.7: 4965260 - 4983395 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04243440_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sbk3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001200041.1 NP_001186970.1 3 - 4 385 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002179.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          biological_process
          phosphorylation
          nucleotide binding
          molecular_function
          protein kinase activity
          protein serine/threonine kinase activity
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964282 AK138915.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964282 NM_001200041.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 133609 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 35905 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 834022 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 579919 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 7097 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 112550 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964282 AK138915.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964282 NM_001200041.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01476187 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115548 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01476187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06174860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06299272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06377351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06517642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06519477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06519460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06174856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06332997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06626858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05938112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06332998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06204082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06161140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06600264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06161136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06252596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06252598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06252597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06204083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06517643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24951298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06377345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06204074 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10559700 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559701 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559699 - -
          Affymetrix Mouse Clariom S Array PSR0700019916.mm.1 NM_001200041.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019916.mm.1 NM_001200041.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019916.mm.1 TC0700002179.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019917.mm.1 NM_001200041.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019917.mm.1 NM_001200041.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019917.mm.1 TC0700002179.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105570161 - -
          Illumina mouse-6 v1.0 expression beadchip 105570161 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1236685 - -
          mouse-6 v1.1 (Illumina) GI_38085684-S - -
          Illumina Sentrix mouse-6.v1 bead array 105570161 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105570161 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2539803 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline