| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600012836.mm.1 |
NR_040296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012836.mm.1 |
NR_040296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012836.mm.1 |
TC0600001604.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600012838.mm.1 |
NR_040296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012838.mm.1 |
NR_040296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012838.mm.1 |
TC0600001604.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600012839.mm.1 |
NR_040296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012839.mm.1 |
NR_040296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012839.mm.1 |
TC0600001604.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600012840.mm.1 |
NR_040296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012840.mm.1 |
NR_040296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012840.mm.1 |
TC0600001604.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600012837.mm.1 |
NR_040296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012837.mm.1 |
NR_040296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600012837.mm.1 |
TC0600001604.mm.1 |
MTA TC |