Search
Search

Mir126a| Gene Symbol: Mir126a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029541.1 | 1 | 38 | 68 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 126b | Mir126b | mmu-mir-126b | Anti-sense | Chr. 2 - 26591363 - 26591420 on NCBI Build GRCm38 | |
| Downstream |
1-acylglycerol-3-phosphate O-acyltransferase 2 (lysophosphatidic acid acyltransferase, beta) |
Agpat2 |
2510002J07Rik, AV000834, BSCL, BSCL1, LPAAB, LPAAT-beta |
1.6kb | Anti-sense | Chr. 2 - 26593057 - 26604417 on NCBI Build GRCm38 |
Set Membership: |
|